missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, 99.8%, for analysis, biochemical grade

CAS: 77-86-1 | C4H11NO3 | 121.14 g/mol

$71.21 - $294.57

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC167621000
SDS View Documents
Thermo Scientific Chemicals
167621000
100 g Plastic bottle
Each for $71.21
Only null left
 
AC167620010
SDS View Documents
Thermo Scientific Chemicals
167620010
1 kg Plastic bottle
Each for $294.57
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

77-86-1
121.14
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
C4H11NO3
MFCD00004679
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
CHEBI:9754
C(C(CO)(CO)N)O

Specifications

77-86-1
>168.5°C
10.0 to 11.5 (5% aq. soln.)
Authentic
Plastic bottle
(HOCH2)3CNH2
100 g
15, 9951
Solubility in water: 550g/L (25°C). Other solubilities: 79.1g/L ethylene glycol- 26g/L methanol (25°C), 22g/L 95% ethanol- 14g/L dmf (25°C), 2g/L acetone- 0.5g/L ethyl acetate (25°C), 0.05g/L chloroform- <0.05g/L ccl4 (25°C)
C(C(CO)(CO)N)O
121.14
CHEBI:9754
99.8%
Powder
(40% aq. soln.) (1 cm cell) (10% aq. soln.) (1 cm cell), (10% aq. soln.) (1 cm cell) (40% aq. soln.) (1 cm cell) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm, 0.2 max. at 290nm
Brown to White
219.0°C to 220.0°C (10.0 mmHg)
99.8%
C4H11NO3
MFCD00004679
04, 303
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
Analysis
Tris(hydroxymethyl)aminomethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-064-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only