missing translation for 'onlineSavingsMsg'
Learn More

Tris(hexafluoroisopropyl) Borate 95.0+%, TCI America™

Catalog No. p-7119179
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16451G 1 g
B16455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16451G Supplier TCI America Supplier No. B16451G
Only null left

Chemical Identifiers

CAS 6919-80-8
Molecular Formula C9H3BF18O3
MDL Number MFCD00191615
Synonym Boric Acid Tris(hexafluoroisopropyl) Ester

Specifications

Melting Point 31°C
Color White
Quantity 1 g
Formula Weight 511.90
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Tris(hexafluoroisopropyl) Borate