missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(hexafluoroisopropyl) Borate 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 31°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 511.90 |
| Percent Purity | ≥95.0% (GC,T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Tris(hexafluoroisopropyl) Borate |
Safety and Handling
| TSCA | No |