missing translation for 'onlineSavingsMsg'
Learn More

Tris-EDTA Buffer Solution, BioUltra, For molecular biology, pH 7.4, MilliporeSigma™ Supelco™

Catalog No. p-7193386
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
Catalog No. Quantity
111016054 100 mL
111004698 500 mL
2 options

Catalog No. 111016054

Supplier: Merck Emd Millipore 93302100ML

Only null left

Specifications

MDL Number MFCD00236359
Quantity 100 mL
Synonym TE buffer solution
Grade For molecular biology