missing translation for 'onlineSavingsMsg'
Learn More

Tris-EDTA Buffer Solution, BioUltra, For molecular biology, pH 7.4, MilliporeSigma™ Supelco™

Catalog No. 111016054
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016054 100 mL
111004698 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111016054 Supplier Merck Emd Millipore Supplier No. 93302100ML

Specifications

MDL Number MFCD00236359
Quantity 100 mL
Synonym TE buffer solution
Grade For molecular biology