missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0

Catalog No. AC327345000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC327345000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC327345000 Supplier Thermo Scientific Chemicals Supplier No. 327345000
Only null left

CAS: 38641-82-6 | C14H27N3O11 | 413.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent, pH buffer

Chemical Identifiers

CAS 38641-82-6
Molecular Formula C14H27N3O11
Molecular Weight (g/mol) 413.38
MDL Number MFCD00236359
InChI Key VLEIUWBSEKKKFX-UHFFFAOYSA-N
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, TRIS, Tris buffer, Tromethamine, Tromethane
PubChem CID 16218629
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid; 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color Colorless
pH 7.9 to 8.1 (25°C)
Physical Form Liquid
Quantity 500 mL
Packaging HDPE Bottle
Beilstein 04, 303
Merck Index 14, 9772
Grade Molecular Biology
Chemical Name or Material Tris EDTA buffer, pH 8.0

RUO – Research Use Only