missing translation for 'onlineSavingsMsg'
Learn More

Tris Dodecyl Sulfate 96.0+%, TCI America™

Catalog No. p-7137558
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3071250MG 250 mg
T30711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3071250MG Supplier TCI America Supplier No. T3071250MG
Only null left

Chemical Identifiers

CAS 50996-85-5
Molecular Formula C16H37NO7S
Molecular Weight (g/mol) 387.532
MDL Number MFCD00043277
InChI Key TYTUELFFKLZOTE-UHFFFAOYSA-N
Synonym Lauryl Sulfate Tris Salt, Dodecyl Sulfate Tris(hydroxymethyl)aminomethane Salt
PubChem CID 11703755
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;dodecyl hydrogen sulfate
SMILES CCCCCCCCCCCCOS(=O)(=O)O.C(C(CO)(CO)N)O

Specifications

Color White-Yellow
Quantity 250 mg
Formula Weight 387.53
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris Dodecyl Sulfate [for Biochemical Research]