missing translation for 'onlineSavingsMsg'
Learn More

Tris(dioxa-3,6-heptyl)amine, 95%

Catalog No. p-3779830
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC124851000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC124851000 Supplier Thermo Scientific Chemicals Supplier No. 124851000

CAS: 70384-51-9 | C15H33NO6 | 323.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70384-51-9
Molecular Formula C15H33NO6
Molecular Weight (g/mol) 323.42
MDL Number MFCD00010748
InChI Key XGLVDUUYFKXKPL-UHFFFAOYSA-N
Synonym tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine
PubChem CID 112414
IUPAC Name 2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine
SMILES COCCOCCN(CCOCCOC)CCOCCOC

Specifications

Color Brown to Yellow
Density 1.0100g/mL
Boiling Point 330.0°C
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4466 to 1.4486
Linear Formula (CH3OCH2CH2OCH2CH2)3N
Quantity 100 mL
Fieser 15,356; 16,373
Specific Gravity 1.01
Solubility Information Solubility in water: miscible. Other solubilities: soluble in acetone, ether, toluene and cyclohexane
Formula Weight 323.42
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tris(dioxa-3, 6-heptyl)amine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 274-590-5

RUO – Research Use Only