missing translation for 'onlineSavingsMsg'
Learn More

Tris(dimethylamino)phosphine (may contain precipitate) 96.0+%, TCI America™

Catalog No. p-7136713
Change view
Click to view available options
Quantity:
25 mL
250 mL
Catalog No. Quantity
T131725ML 25 mL
T1317250ML 250 mL
2 options

Catalog No. T131725ML

Supplier: TCI America T131725ML

Chemical Identifiers

CAS 1608-26-0
Molecular Formula C6H18N3P
Molecular Weight (g/mol) 163.21
MDL Number MFCD00008301
InChI Key XVDBWWRIXBMVJV-UHFFFAOYSA-N
Synonym hexamethylphosphorous triamide, tris dimethylamino phosphine, hexametapil, phosphorous triamide, hexamethyl, hexamethyltriamidophosphite, hexamethyltriaminophosphine, phosphine, tris dimethylamino, bis dimethylamino phosphanyl dimethylamine, hmpt van, hexamethyl phosphorus triamide
PubChem CID 15355
IUPAC Name [bis(dimethylamino)phosphanyl]dimethylamine
SMILES CN(C)P(N(C)C)N(C)C

Specifications

Color Yellow
Boiling Point 163°C
Quantity 25 mL
UN Number 1993
Formula Weight 163.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(dimethylamino)phosphine (may contain precipitate)
RTECSNumber TH3390000
TSCA Yes
Recommended Storage Refrigerator