Learn More
Tris(dimethylamino)aluminum dimer
Preparation of aluminum nitrides | CAS: 32093-39-3 | C12H36Al2N6 | 318.425 g/mol
$1075.07 - $1075.07
Chemical Identifiers
| CAS | 32093-39-3 |
|---|---|
| Molecular Formula | C12H36Al2N6 |
| Molecular Weight (g/mol) | 318.425 |
| MDL Number | MFCD00269811 |
| InChI Key | JGZUJELGSMSOID-UHFFFAOYSA-N |
| Synonym | tris dimethylamido aluminum iii, tris dimethylamino aluminum dimer, bis bis dimethylamino alumanyl dimethylamine, bis mu-dimethylamino tetrakis dimethylamino dialuminum |
| PubChem CID | 16688802 |
| IUPAC Name | N-[bis(dimethylamino)alumanyl]-N-methylmethanamine |
| SMILES | CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C |
Description
Preparation of aluminum nitrides
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
| 32093-39-3 | |
| 318.425 | |
| JGZUJELGSMSOID-UHFFFAOYSA-N | |
| 16688802 | |
| CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C |
| C12H36Al2N6 | |
| MFCD00269811 | |
| tris dimethylamido aluminum iii, tris dimethylamino aluminum dimer, bis bis dimethylamino alumanyl dimethylamine, bis mu-dimethylamino tetrakis dimethylamino dialuminum | |
| N-[bis(dimethylamino)alumanyl]-N-methylmethanamine |
Specifications
| 32093-39-3 | |
| Brown to White | |
| 90°C (0.05 mmHg)(sublimation) | |
| C12H36Al2N6 | |
| MFCD00269811 | |
| UN3131 | |
| tris dimethylamido aluminum iii, tris dimethylamino aluminum dimer, bis bis dimethylamino alumanyl dimethylamine, bis mu-dimethylamino tetrakis dimethylamino dialuminum | |
| CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C | |
| 318.425 | |
| 318.42 | |
| Tris (dimethylamino)aluminum dimer |
| 87°C to 89°C | |
| 0.865 g/mL | |
| 21°C (70°F) | |
| Al2[N(CH3)2]6 | |
| 5 g | |
| Moisture sensitive | |
| JGZUJELGSMSOID-UHFFFAOYSA-N | |
| N-[bis(dimethylamino)alumanyl]-N-methylmethanamine | |
| 16688802 | |
| Powder |
Safety and Handling
GHS H Statement
H228-H260-H314-H318
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P223-P231+P232-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H228-H260-H314-H335
DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
TSCA : No
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only