missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium(0) 75.0+%, TCI America™

Catalog No. p-7137110
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T21841G 1 g
T21845G 5 g
2 options

Catalog No. T21841G

Supplier: TCI America T21841G

Only null left

Chemical Identifiers

CAS 51364-51-3
Molecular Formula C51H42O3Pd2
Molecular Weight (g/mol) 915.73
MDL Number MFCD00013310
InChI Key CYPYTURSJDMMMP-UHFFFAOYSA-N
Synonym tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladium
PubChem CID 9811564
IUPAC Name tris(1,5-diphenylpenta-1,4-dien-3-one) dipalladium
SMILES [Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1

Specifications

Melting Point 155°C
Quantity 1 g
Formula Weight 915.73
Percent Purity ≥75.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(dibenzylideneacetone)dipalladium(0)