missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(dibenzoylmethanato) Iron 97.0+%, TCI America™
$262.83 - $782.90
Chemical Identifiers
| CAS | 14405-49-3 |
|---|---|
| Molecular Formula | C45H33FeO6 |
| Molecular Weight (g/mol) | 725.60 |
| MDL Number | MFCD00192557,MFCD14636458 |
| InChI Key | JWPHPBLJVLHKOZ-UHFFFAOYSA-K |
| Synonym | tris dibenzoylmethanato iron, iron iii diphenylpropanedionate |
| PubChem CID | 57369570 |
| IUPAC Name | iron(3+) tris(3-oxo-1,3-diphenylprop-1-en-1-olate) |
| SMILES | [Fe+3].[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 14405-49-3 | |
| 725.60 | |
| JWPHPBLJVLHKOZ-UHFFFAOYSA-K | |
| 57369570 | |
| [Fe+3].[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
| C45H33FeO6 | |
| MFCD00192557,MFCD14636458 | |
| tris dibenzoylmethanato iron, iron iii diphenylpropanedionate | |
| iron(3+) tris(3-oxo-1,3-diphenylprop-1-en-1-olate) |
Specifications
| 14405-49-3 | |
| MFCD00192557,MFCD14636458 | |
| tris dibenzoylmethanato iron, iron iii diphenylpropanedionate | |
| [Fe+3].[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 725.60 | |
| 725.60 | |
| Crystalline Powder |
| C45H33FeO6 | |
| 5 g | |
| JWPHPBLJVLHKOZ-UHFFFAOYSA-K | |
| iron(3+) tris(3-oxo-1,3-diphenylprop-1-en-1-olate) | |
| 57369570 | |
| ≥97.0% (T) | |
| Tris(dibenzoylmethanato) Iron |
Safety and Handling
TSCA : No