missing translation for 'onlineSavingsMsg'
Learn More

Tris(cyclopentadienyl)yttrium(III), 99.9% (REO)

Catalog No. p-7049476
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1447802 0.5 g
AA1447804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1447802 Supplier Thermo Scientific Chemicals Supplier No. 01447802
Only null left

CAS: 1294-07-1 | C15H15Y | 284.191 g/mol

Tris(cyclopentadienyl)yttrium(III) is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 1294-07-1
Molecular Formula C15H15Y
Molecular Weight (g/mol) 284.191
MDL Number MFCD00058706
InChI Key IUJOPJIXAVSZIQ-UHFFFAOYSA-N
Synonym yttrium 3+ ion tris cyclopenta-1,3-dien-1-ide
PubChem CID 22481132
IUPAC Name cyclopenta-1,3-diene;yttrium(3+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Y+3]

Specifications

Melting Point 295°C
Boiling Point 200°C (sublimation)(vacuum)
Packaging Ampule
Quantity 0.5 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 284.2
Percent Purity 99.9%
Physical Form Powder
Assay (REO)
Chemical Name or Material Tris(cyclopentadienyl)yttrium(III)
Show More Show Less
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber 215-069-4
TSCA No
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only