missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TRIS-buffered saline (TBS, 20X), with 0.5% Tween 20

Catalog No. p-7042763
Change view
Click to view available options
Quantity:
1L
2L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63682K2 1L
AAJ63682K3 2L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63682K2 Supplier Thermo Scientific™ Supplier No. J63682K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00678198
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 20X
Quantity 1L
Chemical Name or Material TRIS-buffered saline, with 0.5% Tween 20
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only