missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS-buffered saline (TBS, 10X), with 1% Triton™ X-100, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It used in some biochemical techniques that is maintain the pH within a relatively narrow range. Tris-HCl is commonly used to make TBS buffers. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash micro titer plates for western blot experiments and screening methods. it is suitable for use as a washing buffer for alkaline phosphatase or peroxidase conjugates in dot blot assay.
Solubility
Fully miscible in water.
Notes
250mM Tris, 27mM KCl, 1.37M NaCl, pH 7.4
It used in some biochemical techniques that is maintain the pH within a relatively narrow range. Tris-HCl is commonly used to make TBS buffers. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash micro titer plates for western blot experiments and screening methods. it is suitable for use as a washing buffer for alkaline phosphatase or peroxidase conjugates in dot blot assay.
Solubility
Fully miscible in water.
Notes
250mM Tris, 27mM KCl, 1.37M NaCl, pH 7.4
Specifications
Specifications
| Concentration | 10X |
| Quantity | 500mL |
| Solubility Information | Fully miscible in water. |
| Chemical Name or Material | TRIS-buffered saline, with 1% Triton™ X-100 |
Safety and Handling
| Hazard Category | H319 |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only