missing translation for 'onlineSavingsMsg'
Learn More

Tris(acetonitrile)cyclopentadienylruthenium(II) Hexafluorophosphate 98.0+%, TCI America™

Catalog No. T3079200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3079200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3079200MG Supplier TCI America Supplier No. T3079200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80049-61-2
Molecular Formula C11H14F6N3PRu
MDL Number MFCD02684569

Specifications

Melting Point 149°C
Quantity 200 mg
Formula Weight 434.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(acetonitrile)cyclopentadienylruthenium(II) Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator