missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, 99 to 99.5%, MP Biomedicals™
Specifications
Specifications
| Color | White |
| Assay Percent Range | 99 to 99.5% |
| Quantity | 10 kg |
| Formula Weight | 121.14 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tris |