missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)
CAS: 17631-68-4 | C30H33EuF21O6 | 1040.52 g/mol
Supplier: Thermo Scientific Chemicals 03354103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III) | |
| 203°C to 207°C | |
| MFCD00064655,MFCD01074933 | |
| Hygroscopic | |
| Soluble in organic solvents. Insoluble in water. | |
| [Eu].CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F | |
| 1040.52 | |
| 1037.49 |
| 17631-68-4 | |
| C30H33EuF21O6 | |
| 1 g | |
| eu fod 3, sievers' reagent, europium-fod, siever's reagent, tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato europium iii, resolve-al tm eufod, chloride ionophore vii, tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato europium, europium iii-tris 1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionate | |
| UDXLMYFGTHAWDC-UHFFFAOYSA-N | |
| europium;(Z)-1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one | |
| 6000041 | |
| Powder |
Chemical Identifiers
| 17631-68-4 | |
| 1040.52 | |
| UDXLMYFGTHAWDC-UHFFFAOYSA-N | |
| 6000041 | |
| [Eu].CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F |
| C30H33EuF21O6 | |
| MFCD00064655,MFCD01074933 | |
| eu fod 3, sievers' reagent, europium-fod, siever's reagent, tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato europium iii, resolve-al tm eufod, chloride ionophore vii, tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato europium, europium iii-tris 1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionate | |
| europium;(Z)-1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one |
Safety and Handling
EINECSNumber : 241-616-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only