missing translation for 'onlineSavingsMsg'
Learn More

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)

Catalog No. AA33541MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33541MD 250 mg
AA3354103 1 g
AA3354106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA33541MD Supplier Thermo Scientific Chemicals Supplier No. 033541MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17631-68-4 | C30H33EuF21O6 | 1040.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17631-68-4
Molecular Formula C30H33EuF21O6
Molecular Weight (g/mol) 1040.52
MDL Number MFCD00064655,MFCD01074933
InChI Key UDXLMYFGTHAWDC-UHFFFAOYSA-N
Synonym eu fod 3, sievers' reagent, europium-fod, siever's reagent, tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato europium iii, resolve-al tm eufod, chloride ionophore vii, tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato europium, europium iii-tris 1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionate
PubChem CID 6000041
IUPAC Name europium;(Z)-1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one
SMILES [Eu].CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 203°C to 207°C
Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 1037.49
Physical Form Powder
Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)
EINECSNumber 241-616-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only