missing translation for 'onlineSavingsMsg'
Learn More

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)erbium(III), 97+%, Thermo Scientific™

Catalog No. p-7051691
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4178302 0.5 g
AA4178304 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4178302 Supplier Thermo Scientific Chemicals Supplier No. 4178302

NMR shift reagent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 17978-75-5
Molecular Formula C30H33ErF21O6
Molecular Weight (g/mol) 1055.81
MDL Number MFCD00074990
InChI Key LEHFMLQRGJCLOZ-UHFFFAOYSA-N
Synonym resolve-al? erfod

Specifications

Melting Point 157°C to 163°C
Quantity 0.5 g
Sensitivity Hygroscopic
Formula Weight 1052.8
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)erbium(III)
TSCA No
Recommended Storage Ambient temperatures