missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-formylphenyl)amine 97.0+%, TCI America™

Catalog No. T2310500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2310500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2310500MG Supplier TCI America Supplier No. T2310500MG
Only null left

Chemical Identifiers

CAS 119001-43-3
Molecular Formula C21H15NO3
Molecular Weight (g/mol) 329.355
InChI Key YOXHQRNDWBRUOL-UHFFFAOYSA-N
Synonym 4,4′C,4′C′C-Nitrilotribenzaldehyde
PubChem CID 231956
IUPAC Name 4-(4-formyl-N-(4-formylphenyl)anilino)benzaldehyde
SMILES C1=CC(=CC=C1C=O)N(C2=CC=C(C=C2)C=O)C3=CC=C(C=C3)C=O

Specifications

Melting Point 243°C
Quantity 500 mg
Formula Weight 329.36
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-formylphenyl)amine