missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-aminophenyl)amine 98.0+%, TCI America™

Catalog No. T23321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23321G 1 g
T23325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23321G Supplier TCI America Supplier No. T23321G

Chemical Identifiers

CAS 5981-09-9
Molecular Formula C18H18N4
Molecular Weight (g/mol) 290.37
MDL Number MFCD00778301
InChI Key SNLFYGIUTYKKOE-UHFFFAOYSA-N
Synonym tris 4-aminophenyl amine, 4,4',4-triaminotriphenylamine, n1,n1-bis 4-aminophenyl benzene-1,4-diamine, 1,4-benzenediamine, n,n-bis 4-aminophenyl, n,n-bis 4-aminophenyl-1,4-benzenediamine, 1,4-benzenediamine, n1,n1-bis 4-aminophenyl, 1-n,1-n-bis 4-aminophenyl benzene-1,4-diamine, pubchem22505, tri 4-aminophenyl amine, tris 4-aminophenyl-amine
PubChem CID 80083
IUPAC Name 4-N,4-N-bis(4-aminophenyl)benzene-1,4-diamine
SMILES C1=CC(=CC=C1N)N(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N

Specifications

Melting Point 230°C
Quantity 1 g
Formula Weight 290.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-aminophenyl)amine