missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-aminophenyl)amine, 97%

Catalog No. p-7041680
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63963MD 250 mg
AAH6396303 1 g
AAH6396306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63963MD Supplier Thermo Scientific Chemicals Supplier No. H63963MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5981-09-9 | C18H18N4 | 290.37 g/mol

A fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 5981-09-9
Molecular Formula C18H18N4
Molecular Weight (g/mol) 290.37
MDL Number MFCD00778301
InChI Key SNLFYGIUTYKKOE-UHFFFAOYSA-N
Synonym tris 4-aminophenyl amine, 4,4',4-triaminotriphenylamine, n1,n1-bis 4-aminophenyl benzene-1,4-diamine, 1,4-benzenediamine, n,n-bis 4-aminophenyl, n,n-bis 4-aminophenyl-1,4-benzenediamine, 1,4-benzenediamine, n1,n1-bis 4-aminophenyl, 1-n,1-n-bis 4-aminophenyl benzene-1,4-diamine, pubchem22505, tri 4-aminophenyl amine, tris 4-aminophenyl-amine
PubChem CID 80083
IUPAC Name 4-N,4-N-bis(4-aminophenyl)benzene-1,4-diamine
SMILES C1=CC(=CC=C1N)N(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N

Specifications

Melting Point 230°C
Quantity 250 mg
Beilstein 2812659
Solubility Information Slightly soluble in water.
Formula Weight 290.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Tris(4-aminophenyl)amine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-791-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only