missing translation for 'onlineSavingsMsg'
Learn More

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride

Catalog No. p-7059128
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4412302 0.5 g
AA4412304 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4412302 Supplier Thermo Scientific Chemicals Supplier No. 04412302

Redox indicator, oxygen sensor | CAS: 36309-88-3 | C72H48Cl2N6Ru | 1169.188 g/mol

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride is used as a probe for luminescent detection and quantitation of oxygen. It is also used for fiber optic sensors. Further, it is used in studies on oxygen in skin and skin tumors as well as measurement of oxygen flux through the skin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36309-88-3
Molecular Formula C72H48Cl2N6Ru
Molecular Weight (g/mol) 1169.188
MDL Number MFCD03095387
InChI Key SKZWFYFFTOHWQP-UHFFFAOYSA-L
Synonym bathophenanthroline ruthenium chloride, ru ddp, tris-bathophenanthroline ruthenium ii chloride, dichlororuthenium-4,7-diphenyl-1,10-phenanthroline 1/3, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride, tris 4,7-diphenyl-1,10-phenanthroline ; dichlororuthenium, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, for fluorescence
PubChem CID 15787787
IUPAC Name dichlororuthenium;4,7-diphenyl-1,10-phenanthroline
SMILES C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.Cl[Ru]Cl

Specifications

Quantity 0.5 g
Solubility Information Soluble in alcohol and dimethylformamide. Insoluble in water.
Formula Weight 1169.2
Physical Form Powder
Chemical Name or Material Tris (4,7-diphenyl-1,10-phenanthroline)ruthenium (II) dichloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only