missing translation for 'onlineSavingsMsg'
Learn More

Tris[4-(5-phenylthiophen-2-yl)phenyl]amine 98.0+%, TCI America™

Catalog No. T3328200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3328200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3328200MG Supplier TCI America Supplier No. T3328200MG
Only null left

Chemical Identifiers

CAS 803727-09-5
Molecular Formula C48H33NS3
Molecular Weight (g/mol) 719.979
InChI Key UJZRJDHTDMTGAP-UHFFFAOYSA-N
PubChem CID 59601002
IUPAC Name 4-(5-phenylthiophen-2-yl)-N,N-bis[4-(5-phenylthiophen-2-yl)phenyl]aniline
SMILES C1=CC=C(C=C1)C2=CC=C(S2)C3=CC=C(C=C3)N(C4=CC=C(C=C4)C5=CC=C(S5)C6=CC=CC=C6)C7=CC=C(C=C7)C8=CC=C(S8)C9=CC=CC=C9

Specifications

Melting Point 213°C
Quantity 200 mg
Formula Weight 719.98
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris[4-(5-phenylthiophen-2-yl)phenyl]amine