missing translation for 'onlineSavingsMsg'
Learn More

Tris[4-(5-bromothiophen-2-yl)phenyl]amine 98.0+%, TCI America™

Catalog No. T3315200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3315200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3315200MG Supplier TCI America Supplier No. T3315200MG

Chemical Identifiers

CAS 339985-36-3
Molecular Formula C30H18Br3NS3
Molecular Weight (g/mol) 728.373
InChI Key QWJSFRTXYYGQPZ-UHFFFAOYSA-N
Synonym 4,4′C,4′C′C-Tris(5-bromothiophen-2-yl)triphenylamine
PubChem CID 91972129
IUPAC Name 4-(5-bromothiophen-2-yl)-N,N-bis[4-(5-bromothiophen-2-yl)phenyl]aniline
SMILES C1=CC(=CC=C1C2=CC=C(S2)Br)N(C3=CC=C(C=C3)C4=CC=C(S4)Br)C5=CC=C(C=C5)C6=CC=C(S6)Br

Specifications

Melting Point 195°C
Color Yellow
Quantity 200 mg
Formula Weight 728.37
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Tris[4-(5-bromothiophen-2-yl)phenyl]amine