missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(3-hydroxypropyltriazolylmethyl)amine 97.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 434.51 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tris(3-hydroxypropyltriazolylmethyl)amine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |