missing translation for 'onlineSavingsMsg'
Learn More

Tris(3,6-dioxaheptyl)amine, 95%

Catalog No. p-7046232
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1354406 5 g
AAL1354414 25 g
AAL1354422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1354406 Supplier Thermo Scientific Chemicals Supplier No. L1354406
Only null left

CAS: 70384-51-9 | C15H33NO6 | 323.43 g/mol

Tris(3,6-dioxaheptyl)amine acts as a phase transfer catalyst used in the preparation of 1,3-dideaza-2'-deoxyadenosine and related benzimidazole 2'-deoxyribonucleosides. It is also used as an intermediate in the synthesis of telithromycin. Further, it is used in the oxidation of arylmethanols under phase-transfer conditions. In addition to this, it acts as a cryptand during phase-transfer glycosylation of 2-amino-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(3,6-dioxaheptyl)amine acts as a phase transfer catalyst used in the preparation of 1,3-dideaza-2′-deoxyadenosine and related benzimidazole 2′-deoxyribonucleosides. It is also used as an intermediate in the synthesis of telithromycin. Further, it is used in the oxidation of arylmethanols under phase-transfer conditions. In addition to this, it acts as a cryptand during phase-transfer glycosylation of 2-amino-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine.

Solubility
Miscible with water.

Notes
Air sensitive and hygroscopic. Incompatible strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 70384-51-9
Molecular Formula C15H33NO6
Molecular Weight (g/mol) 323.43
MDL Number MFCD00010748
InChI Key XGLVDUUYFKXKPL-UHFFFAOYSA-N
Synonym tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine
PubChem CID 112414
IUPAC Name 2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine
SMILES COCCOCCN(CCOCCOC)CCOCCOC

Specifications

Melting Point <-100°C
Density 1.011
Boiling Point 163°C to 171°C (1 mmHg)
Flash Point 162°C (323°F)
Refractive Index 1.4486
Quantity 5 g
Beilstein 2369296
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 323.43
Percent Purity 95%
Chemical Name or Material Tris(3,6-dioxaheptyl)amine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 274-590-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only