missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-methoxyphenyl)phosphine, 97+%, Thermo Scientific™

Chemical Identifiers

CAS 4731-65-1
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00014892
InChI Key IIOSDXGZLBPOHD-UHFFFAOYSA-N
Synonym tris 2-methoxyphenyl phosphine, tris o-methoxyphenyl phosphine, tris 2-methoxyphenyl phosphane, phosphine, tris methoxyphenyl, tris-2-methoxyphenyl-phosphine, tris o-anisyl phosphine, tri-2-methoxyphenyl phosphine, pubchem19707, tri 2-methoxyphenyl phosphine, ksc286k3b
PubChem CID 78464
IUPAC Name tris(2-methoxyphenyl)phosphane
SMILES COC1=CC=CC=C1P(C2=CC=CC=C2OC)C3=CC=CC=C3OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC378850010
SDS View Documents
Thermo Scientific Chemicals
378850010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC378850050
SDS View Documents
Thermo Scientific Chemicals
378850050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4731-65-1
352.37
IIOSDXGZLBPOHD-UHFFFAOYSA-N
78464
COC1=CC=CC=C1P(C2=CC=CC=C2OC)C3=CC=CC=C3OC
C21H21O3P
MFCD00014892
tris 2-methoxyphenyl phosphine, tris o-methoxyphenyl phosphine, tris 2-methoxyphenyl phosphane, phosphine, tris methoxyphenyl, tris-2-methoxyphenyl-phosphine, tris o-anisyl phosphine, tri-2-methoxyphenyl phosphine, pubchem19707, tri 2-methoxyphenyl phosphine, ksc286k3b
tris(2-methoxyphenyl)phosphane

Specifications

4731-65-1
97% min. (HPLC)
C21H21O3P
1g
Solubility in water: insoluble.
COC1=CC=CC=C1P(C2=CC=CC=C2OC)C3=CC=CC=C3OC
352.37
352.37
Tris(2-methoxyphenyl)phosphine
Authentic
Glass bottle
MFCD00014892
tris 2-methoxyphenyl phosphine, tris o-methoxyphenyl phosphine, tris 2-methoxyphenyl phosphane, phosphine, tris methoxyphenyl, tris-2-methoxyphenyl-phosphine, tris o-anisyl phosphine, tri-2-methoxyphenyl phosphine, pubchem19707, tri 2-methoxyphenyl phosphine, ksc286k3b
IIOSDXGZLBPOHD-UHFFFAOYSA-N
tris(2-methoxyphenyl)phosphane
78464
97+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-235-8

SDS
Documents

Documents

RUO – Research Use Only