missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-ethylhexyl) Trimellitate 97.0+%, TCI America™

Catalog No. B088125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B088125ML 25 mL
B0881500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B088125ML Supplier TCI America Supplier No. B088125ML
Only null left

Chemical Identifiers

CAS 3319-31-1
Molecular Formula C33H54O6
Molecular Weight (g/mol) 546.789
MDL Number MFCD00053799
InChI Key KRADHMIOFJQKEZ-UHFFFAOYSA-N
Synonym totm, tris 2-ethylhexyl trimellitate, kodaflex totm, tri-2-ethylhexyl trimellitate, staflex totm, morflex 510, 1,2,4-benzenetricarboxylic acid, tris 2-ethylhexyl ester, hatcol 200, trimex t 08
PubChem CID 18725
IUPAC Name tris(2-ethylhexyl) benzene-1,2,4-tricarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC(=C(C=C1)C(=O)OCC(CC)CCCC)C(=O)OCC(CC)CCCC

Specifications

Color Yellow
Boiling Point 414°C
Quantity 25 mL
Formula Weight 546.79
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-ethylhexyl) Trimellitate
EINECSNumber (3)-1372&(3)-2684
RTECSNumber DC2080000
TSCA Yes