missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-ethylhexyl) Phosphite 90.0+%, TCI America™

Supplier:  TCI America P116925ML

 View more versions of this product

Catalog No. P116925ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tris(2-ethylhexyl) Phosphite
Yellow
MFCD00015266
Phosphorous Acid Trioctyl Ester, Phosphorous Acid Tris(2-ethylhexyl) Ester, Trioctyl Phosphite
CCCCC(CC)COP(OCC(CC)CCCC)OCC(CC)CCCC
418.643
418.64
Liquid
301-13-3
C24H51O3P
25 mL
ILLOBGFGKYTZRO-UHFFFAOYSA-N
tris(2-ethylhexyl) phosphite
9320
≥90.0% (GC)

Chemical Identifiers

301-13-3
418.643
ILLOBGFGKYTZRO-UHFFFAOYSA-N
9320
CCCCC(CC)COP(OCC(CC)CCCC)OCC(CC)CCCC
C24H51O3P
MFCD00015266
Phosphorous Acid Trioctyl Ester, Phosphorous Acid Tris(2-ethylhexyl) Ester, Trioctyl Phosphite
tris(2-ethylhexyl) phosphite
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TH2090000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions