missing translation for 'onlineSavingsMsg'
Learn More

Tris[2-(dimethylamino)ethyl]amine 98.0+%, TCI America™

Catalog No. T28985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28985G 5 g
T28981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28985G Supplier TCI America Supplier No. T28985G

Chemical Identifiers

CAS 33527-91-2
Molecular Formula C12H30N4
Molecular Weight (g/mol) 230.40
MDL Number MFCD00015607
InChI Key VMGSQCIDWAUGLQ-UHFFFAOYSA-N
Synonym n1,n1-bis 2-dimethylamino ethyl-n2,n2-dimethylethane-1,2-diamine, tris 2-dimethylaminoethyl amine, tris 2-dimethylamino ethyl amine, me6tren, 1,2-ethanediamine, n,n-bis 2-dimethylamino ethyl-n',n'-dimethyl, 2-bis 2-dimethylamino ethyl amino ethyl dimethylamine, acmc-20ah4b, n,n-bis 2-dimethylamino ethyl-n',n'-dimethylethane-1,2-diamine
PubChem CID 263094
IUPAC Name (2-{bis[2-(dimethylamino)ethyl]amino}ethyl)dimethylamine
SMILES CN(C)CCN(CCN(C)C)CCN(C)C

Specifications

Color Yellow
Boiling Point 71°C
Quantity 5 g
UN Number 2735
Formula Weight 230.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris[2-(dimethylamino)ethyl]amine
EINECSNumber (7)-0005
TSCA Yes