missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-carboxyethyl)phosphine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America T165625G
Specifications
| Tris(2-carboxyethyl)phosphine Hydrochloride | |
| 177°C | |
| C9H12O6P | |
| 25 g | |
| PZBFGYYEXUXCOF-UHFFFAOYSA-K | |
| 3-[bis(2-carboxylatoethyl)phosphanyl]propanoate | |
| 2734570 | |
| ≥98.0% (T) |
| 51805-45-9 | |
| White | |
| MFCD00145469 | |
| tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride | |
| [O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O | |
| 247.16 | |
| 286.64 | |
| Crystalline Powder |
Chemical Identifiers
| 51805-45-9 | |
| 247.16 | |
| PZBFGYYEXUXCOF-UHFFFAOYSA-K | |
| 2734570 | |
| [O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O |
| C9H12O6P | |
| MFCD00145469 | |
| tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride | |
| 3-[bis(2-carboxylatoethyl)phosphanyl]propanoate |
Safety and Handling
TSCA : No