missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Tris(2-Carboxyethyl)Phosphine Hydrochloride

Numéro de catalogue. C470650G
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C470650G 10 g
C47062G 50 g
C470610G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Numéro de catalogue. C470650G Fournisseur Sigma Aldrich Code fournisseur C470650G
Il en reste null
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(2-carboxyethyl)phosphine hydrochloride (TCEP.HCl) is a non-volatile solid. It is a strong reducing agent.1 It can be synthesized by the acid hydrolysis of tris(2-cyanoethyl)phosphine in refluxing aqueous HCl.1 It has various biological applications such as in vitro and in vivo reduction of disulfide bonds in various peptides and proteins. TCEP is a useful chelating agent for various heavy metal ions as Zn(II), Cd(II), Pb(II), and Ni(II).Water soluble reagent, used for selective reduction of disulfides. More stable than DTT and useful in mass spectrometry applications.

Spécifications

CAS 51805-45-9
Molecular Formula C9H15O6P -+ HCl
Linear Formula C9H15O6P -+ HCl
MDL Number MFCD00145469
Quantity 10 g
Synonym TCEP
Molecular Weight (g/mol) 286.65 g/mol