missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Tris(2-Carboxyethyl)Phosphine Hydrochloride

Catalog No. C470610G
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C470610G 500 g
C47062G 50 g
C470650G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. C470610G Supplier Sigma Aldrich Supplier No. C470610G
Only null left
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(2-carboxyethyl)phosphine hydrochloride (TCEP.HCl) is a non-volatile solid. It is a strong reducing agent.1 It can be synthesized by the acid hydrolysis of tris(2-cyanoethyl)phosphine in refluxing aqueous HCl.1 It has various biological applications such as in vitro and in vivo reduction of disulfide bonds in various peptides and proteins. TCEP is a useful chelating agent for various heavy metal ions as Zn(II), Cd(II), Pb(II), and Ni(II).Water soluble reagent, used for selective reduction of disulfides. More stable than DTT and useful in mass spectrometry applications.

Specifications

CAS 7585-39-9
Melting Point 290°C to 300°C
Molecular Formula C42H70O35
Linear Formula C42H70O35
Quantity 500 g
MDL Number MFCD00078139
Molecular Weight (g/mol) 1134.98 g/mol