missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-aminoethyl)amine, 98+%, Thermo Scientific™

Catalog No. AC314795000
Change view
Click to view available options
500mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC314795000 500mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC314795000 Supplier Thermo Scientific Chemicals Supplier No. 314795000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 4097-89-6
Density 0.9770g/mL
Boiling Point 265.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C6H18N4
Refractive Index 1.4960 to 1.4980
Linear Formula (H2NCH2CH2)3N
Quantity 500mL
Beilstein 04,II,695
Specific Gravity 0.977
Synonym tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine
Solubility Information Solubility in water: soluble.
InChI Key MBYLVOKEDDQJDY-UHFFFAOYSA-N
SMILES C(CN(CCN)CCN)N
IUPAC Name N',N'-bis(2-aminoethyl)ethane-1,2-diamine
Molecular Weight (g/mol) 146.24
PubChem CID 77731
ChEBI CHEBI:30631
Formula Weight 146.24
Percent Purity 96%
Chemical Name or Material Tris(2-aminoethyl)amine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
EINECSNumber 223-857-4

RUO – Research Use Only