missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-pentanedionato)ruthenium(III), TCI America™

Catalog No. T21831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21831G 1 g
T21835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T21831G Supplier TCI America Supplier No. T21831G
Only null left

Chemical Identifiers

CAS 14284-93-6
Molecular Formula C15H21O6Ru
Molecular Weight (g/mol) 398.40
MDL Number MFCD00000030
InChI Key RTZYCRSRNSTRGC-LNTINUHCSA-K
Synonym ruthenium acetylacetonate, ruthenium iii acetylacetonate, acetylacetone ruthenium iii salt, tris 2,4-pentanedionato ruthenium iii, tris pentane-2,4-dionato-o,o' ruthenium, ruthenium, tris 2,4-pentanedionato-o,o'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-kappao,kappao'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-8ci, z-4-oxopent-2-en-2-olate; ruthenium 3+
PubChem CID 5488829
IUPAC Name ruthenium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Ru+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 260°C
Color Red
Quantity 1 g
Formula Weight 398.40
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)ruthenium(III)