missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-pentanedionato)lanthanum(III) Hydrate 98.0+%, TCI America™

Catalog No. L014825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L014825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L014825G Supplier TCI America Supplier No. L014825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity rare-earth metal compound for chemical research applications.

  • Laboratory reagent to be stored in room temperature in a cool and dark place

Chemical Identifiers

CAS 64424-12-0
Molecular Formula C15H21LaO6
Molecular Weight (g/mol) 436.23
MDL Number MFCD00149057
InChI Key UUYNRMJIFVOQCR-UHFFFAOYSA-N
Synonym lanthanum acetylacetonate hydrate min
PubChem CID 132984033
IUPAC Name lanthanum(3+) tris(2,4-dioxopentan-3-ide)
SMILES [La+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range >98.0%
Solubility Information Insoluble in Water
Formula Weight 436.23
Percent Purity ≥98.0% (T)
Appearance White to Light Yellow to Light Orange Powder to Crystal
Chemical Name or Material Tris(2,4-pentanedionato)lanthanum(III) Hydrate