missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-pentanedionato)iron(III) 98.0+%, TCI America™

Catalog No. I0079500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0079500G 500 g
I007925G 25 g
I0079100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0079500G Supplier TCI America Supplier No. I0079500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14024-18-1
Molecular Formula C15H21FeO6
Molecular Weight (g/mol) 353.17
MDL Number MFCD00000020
InChI Key AQBLLJNPHDIAPN-LNTINUHCSA-K
Synonym Acetylacetone Iron(III) Salt, Ferric(III) Acetylacetonate, Iron(III) Acetylacetonate
IUPAC Name iron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 185°C
Color Red
Quantity 500 g
Formula Weight 353.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)iron(III)
EINECSNumber (2)-2142
RTECSNumber NO8960000
TSCA Yes