missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-pentanedionato)indium(III) 99.0+%, TCI America™

Catalog No. I002010G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I002010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I002010G Supplier TCI America Supplier No. I002010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14405-45-9
Molecular Formula C15H21InO6
Molecular Weight (g/mol) 412.15
MDL Number MFCD00013494
InChI Key CMGVNFXMZKDHDG-UHFFFAOYSA-N
Synonym acetylacetone indium iii salt, indium iii tris 4-oxo-2-pentene-2-olate
PubChem CID 101644361
IUPAC Name indium(3+) tris(2,4-dioxopentan-3-ide)
SMILES [In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 10 g
Formula Weight 412.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)indium(III)
RTECSNumber NL2025000
TSCA Yes