missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2,4-pentanedionato)gallium(III) 97.0+%, TCI America™
$140.54 - $475.52
Chemical Identifiers
| CAS | 14405-43-7 |
|---|---|
| Molecular Formula | C15H21GaO6 |
| Molecular Weight (g/mol) | 367.05 |
| MDL Number | MFCD00013492 |
| InChI Key | IMMRYVXLCAMRNK-UHFFFAOYSA-N |
| Synonym | gallium iii acetylacetonate, gallium iii 2,4-pentanedionate |
| PubChem CID | 71311502 |
| IUPAC Name | gallium(3+) tris(2,4-dioxopentan-3-ide) |
| SMILES | [Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Chemical Identifiers
| 14405-43-7 | |
| 367.05 | |
| IMMRYVXLCAMRNK-UHFFFAOYSA-N | |
| 71311502 | |
| [Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
| C15H21GaO6 | |
| MFCD00013492 | |
| gallium iii acetylacetonate, gallium iii 2,4-pentanedionate | |
| gallium(3+) tris(2,4-dioxopentan-3-ide) |
Specifications
| 14405-43-7 | |
| White-Yellow | |
| MFCD00013492 | |
| gallium iii acetylacetonate, gallium iii 2,4-pentanedionate | |
| [Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
| 367.05 | |
| 367.05 | |
| Crystalline Powder |
| 200°C | |
| C15H21GaO6 | |
| 5 g | |
| IMMRYVXLCAMRNK-UHFFFAOYSA-N | |
| gallium(3+) tris(2,4-dioxopentan-3-ide) | |
| 71311502 | |
| ≥97.0% (T) | |
| Tris(2,4-pentanedionato)gallium(III) |
Safety and Handling
TSCA : No