missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-pentanedionato)cobalt(III) 98.0+%, TCI America™

Catalog No. p-7124382
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
C037425G 25 g
C0374100G 100 g
C0374500G 500 g
3 options

Catalog No. C037425G

Supplier: TCI America C037425G

Only null left

Chemical Identifiers

CAS 21679-46-9
Molecular Formula C15H21CoO6
Molecular Weight (g/mol) 356.26
MDL Number MFCD00013488
InChI Key VACIXPOMJJIDBM-UHFFFAOYSA-N
Synonym Acetylacetone Cobalt(III) Salt, Cobalt(III) Acetylacetonate
IUPAC Name 2,4,8,10,13,15-hexamethyl-1λ³,5λ³,7λ³,11λ³,12λ³,16λ³-hexaoxa-6-cobaltaspiro[5.5⁶.5⁶]hexadeca-1,4,7,10,12,15-hexaene-6,6,6-tris(ylium)-3,9,14-triide
SMILES CC1=[O][Co+3]234([O]=C(C)[CH-]1)[O]=C(C)[CH-]C(C)=[O]2.CC([CH-]C(C)=[O]3)=[O]4

Specifications

Quantity 25 g
Formula Weight 356.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)cobalt(III)
EINECSNumber (2)-2142
TSCA Yes