missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,3-dibromopropyl) isocyanurate, 97%, Thermo Scientific Chemicals

Catalog No. AAH6005636
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6005636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6005636 Supplier Thermo Scientific Chemicals Supplier No. H6005636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52434-90-9 | Molecular Formula: C12H15Br6N3O3 | Molecular Weight: 728.694 g/mol

Tris(2,3-dibromopropyl) isocyanurate is a non degradable heterocyclic hexabrominated flame retardant. It inhibited the expression of psaA and psbC that led to the decrease of Nannochloropsis sp.s growth biomass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2,3-dibromopropyl) isocyanurate is a non degradable heterocyclic hexabrominated flame retardant. It inhibited the expression of psaA and psbC that led to the decrease of Nannochloropsis sp.′s growth biomass.

Solubility
Soluble in chloroform.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 52434-90-9
Molecular Formula C12H15Br6N3O3
Molecular Weight (g/mol) 728.694
MDL Number MFCD00010290
InChI Key NZUPFZNVGSWLQC-UHFFFAOYSA-N
Synonym tris 2,3-dibromopropyl isocyanurate, 1,3,5-tris 2,3-dibromopropyl-1,3,5-triazinane-2,4,6-trione, hexahydro-1,3,5-tris 2,3-dibromopropyl-1,3,5-triazine-2,4,6-trione, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3,5-tris 2,3-dibromopropyl, 1,3,5-tris 2,3-dibromopropyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione, isocyanuric acid tris 2,3-dibromopropyl ester, 1,3,5-tris 2,3-dibromopropyl-s-triazine-2,4,6-trione, acmc-1apwt, tri 2,3-dibromopropyl isocyanate
PubChem CID 103634
IUPAC Name 1,3,5-tris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione
SMILES C(C(CBr)Br)N1C(=O)N(C(=O)N(C1=O)CC(CBr)Br)CC(CBr)Br

Specifications

Quantity 500 g
Solubility Information Soluble in chloroform.
Formula Weight 728.69
Percent Purity 97%
Chemical Name or Material Tris(2,3-dibromopropyl) isocyanurate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 257-913-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only