missing translation for 'onlineSavingsMsg'
Learn More

Tris[2-(2-methoxyethoxy)ethyl]amine 90.0+%, TCI America™

Catalog No. p-7136670
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T123125G 25 g
T1231100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T123125G Supplier TCI America Supplier No. T123125G

Chemical Identifiers

CAS 70384-51-9
Molecular Formula C15H33NO6
Molecular Weight (g/mol) 323.43
MDL Number MFCD00010748
InChI Key XGLVDUUYFKXKPL-UHFFFAOYSA-N
Synonym tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine
PubChem CID 112414
IUPAC Name 2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine
SMILES COCCOCCN(CCOCCOC)CCOCCOC

Specifications

Boiling Point 171°C
Quantity 25 g
UN Number 2735
Formula Weight 323.43
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris[2-(2-methoxyethoxy)ethyl]amine