missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2'-bipyridyl)ruthenium(II) Chloride Hexahydrate 98.0+%, TCI America™

Catalog No. p-7136889
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16551G 1 g
T16555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16551G Supplier TCI America Supplier No. T16551G

Chemical Identifiers

CAS 50525-27-4
Molecular Formula C30H36Cl2N6O6Ru
Molecular Weight (g/mol) 748.62
MDL Number MFCD00149670
InChI Key WHELTKFSBJNBMQ-UHFFFAOYSA-L
Synonym tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
PubChem CID 131664161
IUPAC Name λ²-ruthenium(2+) tris(2,2'-bipyridine) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Specifications

Color Red-Yellow
Quantity 1 g
Formula Weight 640.53
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,2′-bipyridyl)ruthenium(II) Chloride Hexahydrate
RTECSNumber VM2730000
TSCA No