missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%

Catalog No. p-3779838
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208750010 1 g Glass bottle
AC208750050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC208750010 Supplier Thermo Scientific Chemicals Supplier No. 208750010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50525-27-4 | C30H36Cl2N6O6Ru | 748.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50525-27-4
Molecular Formula C30H36Cl2N6O6Ru
Molecular Weight (g/mol) 748.62
MDL Number MFCD00149670
InChI Key WHELTKFSBJNBMQ-UHFFFAOYSA-L
Synonym tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
PubChem CID 131664161
IUPAC Name 2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Specifications

Melting Point >300.0°C
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 748.63
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tris(2, 2'-bipyridyl)ruthenium(II) chloride, hexahydrate
Show More Show Less

RUO – Research Use Only