missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2'-bipyridine)cobalt(II) Bis(hexafluorophosphate) 95.0+%, TCI America™

Catalog No. T32551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32551G 1 g
T32555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32551G Supplier TCI America Supplier No. T32551G
Only null left

Chemical Identifiers

CAS 79151-78-3
Molecular Formula C30H24CoF12N6P2
Molecular Weight (g/mol) 817.426
InChI Key IDXASOGRZNYSPC-UHFFFAOYSA-N
PubChem CID 15610156
IUPAC Name cobalt(2+);2-pyridin-2-ylpyridine;dihexafluorophosphate
SMILES C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.[Co+2]

Specifications

Quantity 1 g
Formula Weight 817.43
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,2′-bipyridine)cobalt(II) Bis(hexafluorophosphate)