missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), recrystallized, 99.9% (metals basis)

Catalog No. AA4186206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4186206 5 g
AA4186214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4186206 Supplier Thermo Scientific Chemicals Supplier No. 04186206

CAS: 14876-47-2 | C33H60FeO6 | 608.682 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14876-47-2
Molecular Formula C33H60FeO6
Molecular Weight (g/mol) 608.682
MDL Number MFCD00067465
InChI Key WLITYJBILWOYFF-QFVJJVGWSA-N
Synonym fe tmhd 3
PubChem CID 15251600
IUPAC Name (E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;iron
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Fe]

Specifications

Melting Point 148°C
Color Red
Boiling Point 300°C (decomposition)
Odor Odorless
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 605.66
Percent Purity 99.9%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), Recrystallized
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only