missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III)

Catalog No. p-7080386
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8789803 1 g
AA8789806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8789803 Supplier Thermo Scientific Chemicals Supplier No. 08789803
Only null left

CAS: 15522-71-1 | C33H60EuO6 | 704.801 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15522-71-1
Molecular Formula C33H60EuO6
Molecular Weight (g/mol) 704.801
MDL Number MFCD00000019
InChI Key RHXUZKJNHAMZEP-QFVJJVGWSA-N
Synonym eu dpm 3, tris 2,2,6,6-tetramethyl-3,5-heptanedionato europium, tris 2,2,6,6-tetramethyl-3,5-heptanedionato europium iii, resolve-al tm, europium iii-dpm, eu tmhd 3, europium 2,2,6,6-tetramethyl-3,5-heptanedionate, europium-tris dipivaloylmethane, tris 2,2,6,6-tetramethyl-3,5-heptanedionato europium 3
PubChem CID 10897800
IUPAC Name europium;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Eu]

Specifications

Melting Point 187°C to 189°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 701.78
Physical Form Crystalline
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)europium (III)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-565-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only