missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III) 95.0+%, TCI America™

Catalog No. p-7136688
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12651G 1 g
T12655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12651G Supplier TCI America Supplier No. T12651G
Only null left

Chemical Identifiers

CAS 15522-71-1
Molecular Formula C33H60EuO6
Molecular Weight (g/mol) 704.801
MDL Number MFCD00000019
InChI Key RHXUZKJNHAMZEP-QFVJJVGWSA-N
Synonym eu dpm 3, tris 2,2,6,6-tetramethyl-3,5-heptanedionato europium, tris 2,2,6,6-tetramethyl-3,5-heptanedionato europium iii, resolve-al tm, europium iii-dpm, eu tmhd 3, europium 2,2,6,6-tetramethyl-3,5-heptanedionate, europium-tris dipivaloylmethane, tris 2,2,6,6-tetramethyl-3,5-heptanedionato europium 3
PubChem CID 10897800
IUPAC Name europium;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Eu]

Specifications

Melting Point 190°C
Color Yellow
Quantity 1 g
Formula Weight 701.78
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III) [NMR Shift Reagent]